missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate (Fine White Powder), Fisher BioReagents™

Product Code. p-2363402
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10244683

Brand: Fisher BioReagents BP328500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium Bicarbonate (Fine White Powder)

This sodium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 84.01
Percent Purity 99.7 to 100.3%
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium Bicarbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.