missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | NaHCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4153970 |
Solubility Information | Soluble in water, 50g/L |
Formula Weight | 84.01g/mol |
Percent Purity | ≥99.7% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Sodium Bicarbonate, Reag. Ph. Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'