missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, purum p.a, ≥99.0% (T), Honeywell™

Product Code. 15676800
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point Decomposes before melting
Color Colorless
pH 8.0 to 8.6
Physical Form Powder
Quantity 1 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
Solubility Information 90g/L at 20°C
IUPAC Name sodium hydrogen carbonate
Formula Weight 84.01g/mol
Percent Purity ≥99.0% (T)
Packaging Poly bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'