missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate USP/FCC Powder, Macron Fine Chemicals™

Product Code. 15558414
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 84.01
Grade FCC/USP
Packaging POLYSTORMOR
Chemical Name or Material Sodium Bicarbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'