missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide, 98%, Thermo Scientific Chemicals

43.70 € - 369.00 €

Chemical Identifiers

CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Molecular Weight (g/mol) 183.377
MDL Number MFCD00009835
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium saltShow MoreShow Less
PubChem CID 2724254
IUPAC Name sodium;bis(trimethylsilyl)azanide
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327748
SDS View Documents
Thermo Scientific Alfa Aesar
033505.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11307748
SDS View Documents
Thermo Scientific Alfa Aesar
033505.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11317748
SDS View Documents
Thermo Scientific Alfa Aesar
033505.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Sodium bis(trimethylsilyl)amide is used as a base in organic synthesis as well as to deprotonate other compounds containing weakly acidic O-H, S-H, and N-H bonds. It is used to prepare amines by reacting with alkyl halides. It finds application in certain organic reactions viz. generation of carbenes by dehydrohalogenation of halocarbons and Wittig reagents by deprotonation of phosphonium salts. It accelerates the polymerization of pheneylacetylene in conjunction with rhodium(I) catalysis. Furthermore, it is used for the preparation of lanthanide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H228-H261-H314-H335-H500

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I

missing translation for 'einecsNumber' : 213-983-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium bis(trimethylsilyl)amide, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.