missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuthate(V), 85%, Thermo Scientific Chemicals

Product Code. 10012770
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.96
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Color Yellow to Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 80% min. (Iodometry)
Linear Formula NaBiO3
Merck Index 15,8721
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids
Formula Weight 279.96
Percent Purity 85%
Packaging Glass bottle
Chemical Name or Material Sodium bismuthate(V)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'