missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bisulfate Monohydrate, puriss p.a., ≥99.0% (T), Honeywell™
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
CAS | 10034-88-5 |
Quantity | 500 g |
Molecular Formula | H3NaO5S |
Linear Formula | NaHSO4 · H2O |
MDL Number | MFCD00149210 |
UN Number | NONH for all modes of transport |
Synonym | sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox |
InChI Key | JXHZRQHZVYDRGX-UHFFFAOYSA-M |
SMILES | O.[Na+].OS([O-])(=O)=O |
IUPAC Name | sodium;hydrogen sulfate;hydrate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'