missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bisulfite, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Specifications
Specifications
Quantity | 100 g |
IUPAC Name | sodium;hydrogen sulfite |
Grade | ACS Reagent |
Chemical Name or Material | Sodium Bisulfite |
Safety and Handling
Product Identifier
- Sodium metabisulfite
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium metabisulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction