missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 98+%, powder, Thermo Scientific Chemicals

29.45 € - 238.19 €

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydroborate, SBH
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10213621
View Documents
Thermo Scientific Acros
189300050
5 g Plastic Bottle
29.45 €
5g
Please to purchase this item. Need a web account? Register with us today!
10193231
View Documents
Thermo Scientific Acros
189300100
10 g Plastic Bottle
29.85 €
10g
Please to purchase this item. Need a web account? Register with us today!
10772421
View Documents
Thermo Scientific Acros
189301000
100 g Plastic Bottle
81.41 €
100g
Please to purchase this item. Need a web account? Register with us today!
10695632
View Documents
Thermo Scientific Acros
189305000
500 g Plastic Bottle
238.19 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

16940-66-2
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
BH4Na
MFCD00003518
Sodium tetrahydroborate, SBH
[BH4-].[Na+]

Specifications

360.0°C
White
98% min. (Iodometry)
NaBH4
01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Sodium tetrahydroborate, SBH
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.83
Negligible
1.3
16940-66-2
Powder
BH4Na
MFCD00003518
15, 8727
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
[BH4-].[Na+]
37.83
≥98%
>220°C
Sodium borohydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 241-004-4

missing translation for 'rtecsNumber' : ED3325000

missing translation for 'tsca' : TSCA

Product Identifier
  • Sodium borohydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H301-Toxic if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates

RUO – Research Use Only