Learn More
Sodium borohydride, 98%, Thermo Scientific Chemicals
80.50 € - 919.00 €
Chemical Identifiers
CAS | 16940-66-2 |
---|---|
Molecular Formula | BH4Na |
Molecular Weight (g/mol) | 37.83 |
MDL Number | MFCD00003518 |
InChI Key | YOQDYZUWIQVZSF-UHFFFAOYSA-N |
Synonym | Sodium tetrahydridoborate |
IUPAC Name | sodium boranuide |
SMILES | [BH4-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316768
|
Thermo Scientific Alfa Aesar
013432.22 |
100 g |
80.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336768
|
Thermo Scientific Alfa Aesar
013432.36 |
500 g |
229.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326768
|
Thermo Scientific Alfa Aesar
013432.57 |
4 x 500 g |
919.00 €
2000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in the ammonia borane dehydrogenation. Sodium borohydride is used as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces the aldehyde and keto groups to give the corresponding alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16940-66-2 | |
37.83 | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide |
BH4Na | |
MFCD00003518 | |
Sodium tetrahydridoborate | |
[BH4-].[Na+] |
Specifications
400°C (decomposition) | |
White | |
98% | |
MFCD00003518 | |
Soluble in water. Also soluble in liquid ammonia,amines,pyridine. | |
[BH4-].[Na+] | |
37.83 | |
220°C (428°F) | |
Moisture sensitive | |
1.074 g/cm3 at 20°C |
16940-66-2 | |
Powder | |
BH4Na | |
Sodium tetrahydridoborate | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.83 | |
Odorless | |
White caplets | |
Sodium Borohydride |
Safety and Handling
P201-P202-P223-P231+P232-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H260-H301-H314-H360FD
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: I
missing translation for 'einecsNumber' : 241-004-4
missing translation for 'rtecsNumber' : ED3325000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Sodium borohydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sodium borohydride
RUO – Research Use Only