missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, Technical, SLR, Fisher Chemical™

22.00 € - 88.65 €

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10409190
View Documents
Fisher Chemical
S/2560/45
10 g
22.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10599010
View Documents
Fisher Chemical
S/2560/46
25 g
46.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
10703621
View Documents
Fisher Chemical
S/2560/48
100 g
88.65 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

16940-66-2
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
[BH4-].[Na+]
BH4Na
MFCD00003518
sodium boranuide

Specifications

360°C
White
Powder/Solid
MFCD00003518
[BH4-].[Na+]
37.83
Plastic powder jar
16940-66-2
11
BH4Na
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.83g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium borohydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H301-Toxic if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications