Learn More
Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 448511000
24.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
130.0°C to 135.0°C | |
VenPure™ solution, 12% aq. solution | |
100 mL | |
NaBH4 | |
15,8727 | |
Solubility in water: completely soluble | |
[BH4-].[Na+] | |
37.83 | |
Plastic bottle | |
1.4000g/mL |
Sodium borohydride | |
16940-66-2, 1310-73-2, 67-56-1, 7732-18-5 | |
BH4Na | |
MFCD00003518 | |
SBH, Sodium tetrahydroborate | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.82 | |
1.4 |
Safety and Handling
- Sodium borohydride, 12% aqueous solution, (VenPureTM solution), contains 40% NaOH
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide, Sodium borohydride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only