missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 448515000
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
130.0°C to 135.0°C | |
VenPure™ solution, 12% aq. solution | |
500 mL | |
NaBH4 | |
15,8727 | |
Solubility in water: completely soluble | |
[BH4-].[Na+] | |
37.83 | |
1.4 |
Sodium borohydride | |
16940-66-2, 1310-73-2, 67-56-1, 7732-18-5 | |
BH4Na | |
MFCD00003518 | |
SBH, Sodium tetrahydroborate | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.82 | |
1.4000g/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction