missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99+% (dry wt.), water <1.0%, Thermo Scientific Chemicals

Product Code. 11488833
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

Solubility
Soluble in water, soluble in alcohol.

Notes
It is thermally and chemically stable. Hygroscopic. Sodium bromide is non-flammable and when strongly heated it may emit bromine fumes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Quantity 250 g
Merck Index 14,8594
Solubility Information Soluble in water,soluble in alcohol.
IUPAC Name sodium bromide
Formula Weight 102.89
Percent Purity ≥99%
Refractive Index 1.6412
Sensitivity Hygroscopic
Density 3.203
Assay (dry wt.)
Chemical Name or Material Sodium bromide, Water <1.0%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium bromide
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.