missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, Honeywell Fluka™
BIOPHARMA

Product Code. 15690560
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Quantity 1 kg
Linear Formula NaBr
UN Number NONH for all modes of transport
IUPAC Name sodium;bromide
Formula Weight 102.89g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 1mmHg (806°C)
Chemical Name or Material Sodium Bromide, Meets analytical specification of Ph. Eur.BP,USP
Product Identifier
  • Sodium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'