missing translation for 'onlineSavingsMsg'
Learn More
Sodium carbonate, ACS primary standard, 99.95-100.05% (dried basis), Thermo Scientific Chemicals Product Code.: 11357698

Sodium carbonate, ACS primary standard, 99.95-100.05% (dried basis), Thermo Scientific Chemicals

Product Code. 11357698
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11357698

Brand: Thermo Scientific Alfa Aesar 033377.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Physical Form Granules
Quantity 500 g
Assay Percent Range 99.95 to 100.05%
Beilstein 4154566
Solubility Information Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates.
IUPAC Name disodium;carbonate
Formula Weight 105.99
Grade ACS Reagent
Refractive Index 1.535
Sensitivity Hygroscopic
Density 2.53 g/mL
Chemical Name or Material Sodium carbonate, ACS primary standard
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium carbonate, ACS primary standard, 99.95-100.05% (dried basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.