missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Anhydrous, Extra Pure, SLR, Fisher Chemical™

33.85 € - 190.00 €

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10538070
SDS View Documents
Fisher Chemical
S/2880/53
500 g Plastic powder jar
33.85 €
500g
Please to purchase this item. Need a web account? Register with us today!
10665692
SDS View Documents
Fisher Chemical
S/2880/60
1 kg Plastic powder jar
45.95 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10316200
SDS View Documents
Fisher Chemical
S/2880/62
2.5 kg Plastic powder jar
105.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10162540
SDS View Documents
Fisher Chemical
S/2880/65
5 kg Plastic keg
190.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium Carbonate Anhydrous, Extra Pure, SLR

Specifications

Chemical Identifiers

497-19-8
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
[Na+].[Na+].[O-]C([O-])=O
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377

Specifications

1600°C
497-19-8
11.3
CNa2O3
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
[Na+].[Na+].[O-]C([O-])=O
105.99
CHEBI:29377
854°C
White
Solid
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
105.99g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium carbonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.