missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Anhydrous, Technical, Fisher Chemical

Product Code. 10477840
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Boiling Point 1600°C
Melting Point 854°C
CAS 497-19-8
Color White
pH 11.3
Physical Form Solid
Quantity 2.5 kg
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium;carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
ChEBI CHEBI:29377
Formula Weight 105.99g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.