Learn More
Sodium carbonate decahydrate, 99+%, Thermo Scientific Chemicals
Description
Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.
Solubility
Soluble in water and glycerol. Insoluble in alcohol.
Notes
Air sensitive. Incompatible with strong acids and aluminum. Keep the container tightly closed.

Specifications
Specifications
Melting Point | 32°C to 34°C |
Quantity | 500 g |
Linear Formula | Na2CO3·10H2O |
Merck Index | 14,8596 |
Solubility Information | Soluble in water and glycerol. Insoluble in alcohol. |
Formula Weight | 286.14 (105.99 Anhydrous) |
Percent Purity | ≥99% |
Density | 1.46 |
Chemical Name or Material | Sodium carbonate decahydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10250069 | Certificate of Analysis | 25/03/2024 | A17484.0C,A17484.36,A17484.0E |
10241212 | Certificate of Analysis | 14/06/2022 | A17484.0C,A17484.36,A17484.0E |
10239174 | Certificate of Analysis | 12/04/2022 | A17484.0C,A17484.36,A17484.0E |
10237372 | Certificate of Analysis | 12/04/2022 | A17484.0C,A17484.36,A17484.0E |
10184366 | Certificate of Analysis | 23/11/2021 | A17484.0C,A17484.36,A17484.0E |
Safety and Handling

- Sodium carbonate decahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sodium carbonate decahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.