Learn More
Sodium carbonate monohydrate, ACS, 99.5% min, Thermo Scientific Chemicals
19.40 € - 130.65 €
Chemical Identifiers
CAS | 5968-11-6 |
---|---|
Molecular Formula | CH2Na2O4 |
Molecular Weight (g/mol) | 124.00 |
MDL Number | MFCD00149177 |
InChI Key | MQRJBSHKWOFOGF-UHFFFAOYSA-L |
Synonym | sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop |
PubChem CID | 2735133 |
IUPAC Name | disodium;carbonate;hydrate |
SMILES | C(=O)([O-])[O-].O.[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347948
|
Thermo Scientific Alfa Aesar
036485.22 |
100 g |
19.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357948
|
Thermo Scientific Alfa Aesar
036485.36 |
500 g |
57.69 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337948
|
Thermo Scientific Alfa Aesar
036485.A3 |
2 kg |
130.65 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5968-11-6 | |
124.00 | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
2735133 | |
C(=O)([O-])[O-].O.[Na+].[Na+] |
CH2Na2O4 | |
MFCD00149177 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
disodium;carbonate;hydrate |
Specifications
100°C-H2O | |
Granules | |
CH2Na2O4 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
disodium;carbonate;hydrate | |
2735133 | |
ACS Reagent | |
2.250 g/mL |
5968-11-6 | |
99.5% | |
MFCD00149177 | |
Soluble in water and glycerol. Insoluble in alcohol,acetone,alkyl acetates,benzonitrile and liquid ammonia. | |
C(=O)([O-])[O-].O.[Na+].[Na+] | |
124.00 | |
124.01 (105.99 Anhydrous) | |
Hygroscopic | |
Sodium carbonate monohydrate, For analysis ACS |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H336
missing translation for 'einecsNumber' : 207-838-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.