missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

22.60 € - 51.10 €

Chemical Identifiers

MDL Number 3494
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10489750
View Documents Promotion Details
Fisher Chemical
J/7520/PB15
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10496870
View Documents Promotion Details
Fisher Chemical
J/7520/17
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

3494

Specifications

100°C
Colorless
3494
14mmHg at 20°C
0°C
Liquid
HDPE plastic bottle
Vapor Density: 0.7
Videos
SDS
Documents

Documents

Product Certifications