missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium Caseinate Pract.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Sodium caseinate is the salt of the phosphoprotein class known as caseins
• Caseins are found in animal tissues, especially milk, and they contain all nine essential amino acids
Applications
• Casein can be used in protease enzyme activity studies, in vitro experiments on cell growth and differentiation, or as a blocking solution for western blot
Specifications
Specifications
Color | White to Cream |
pH | 6 to 7.5 (5% soln.) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 100 g |
Percent Purity | ≥92% (protein) (on dry substance) |
Physical Form | Powder |
Chemical Name or Material | Sodium caseinate, pract. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'