missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11495030
Click to view available options
Quantity:
500 mL
1 L

Sodium chloride, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Quantity 500 mL
Color Colorless
IUPAC Name sodium;chloride
Formula Weight 58.44
Concentration or Composition (by Analyte or Components) Sodium chloride: 0.6%; Water: 99.4%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Miscible with water, alcohol and ammonia
Physical Form Liquid
Chemical Name or Material Sodium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'