missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free

67.03 € - 131.66 €

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10297760
View Documents Promotion Details
Thermo Scientific Chemicals
327300010
1 kg
67.03 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10268163
View Documents Promotion Details
Thermo Scientific Chemicals
327300025
2.5 kg
131.66 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]

Specifications

1461.0°C
7647-14-5
Crystalline Solid
ClNa
MFCD00003477
15, 8734
Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
[Na+].[Cl-]
58.44
CHEBI:26710
99.85%
Glass Bottle
801.0°C
White
99.8% min. (Argentometry)
NaCl
13,280
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
Molecular Biology
Sodium chloride, DNAse, RNAse and Protease Free
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-598-3

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only