missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
45.43 € - 1842.69 €
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15658170
|
Honeywell-Fluka
S9888-25G |
25 g | N/A | ||||||
15688170
|
Honeywell-Fluka
S9888-500G |
500 g |
52.54 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15628170
|
Honeywell-Fluka
S9888-1KG |
1 kg |
89.97 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15638170
|
Honeywell-Fluka
S9888-2.5KG |
2.5 kg |
209.84 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15618180
|
Honeywell-Fluka
S9888-6X500G |
6 x 500 g |
267.90 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15608180
|
Honeywell-Fluka
S9888-5KG |
5 kg |
220.66 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15678170
|
Honeywell-Fluka
S9888-4X2.5KG |
4 x 2.5 kg |
534.24 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15618170
|
Honeywell-Fluka
S9888-10KG |
10 kg |
257.83 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15648170
|
Honeywell-Fluka
S9888-20KG |
20 kg |
628.67 €
20kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15668170
|
Honeywell-Fluka
S9888-25KG |
25 kg |
930.65 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15698170
|
Honeywell-Fluka
S9888-50KG |
50 kg |
1842.69 €
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Identifiants chimiques
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Spécification
7647-14-5 | |
NaCl | |
NONH for all modes of transport | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
≥99% | |
Sodium chloride |
ClNa | |
MFCD00003477 | |
3534976 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44g/mol | |
ACS Reagent |
Sécurité et traitement
missing translation for 'rtecsNumber' : VZ4725000