missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Fisher BioReagents™

Product Code. 10123443
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Pail

Sodium Chloride

Sodium Chloride is a main component of many electrophoresis and molecular biology buffers and media.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 10 kg
Assay Percent Range ≥99.0 %
Merck Index 15, 8734
IUPAC Name sodium;chloride
Formula Weight 58.44
Percent Purity ≥99.0%
Identification Pass Test
Packaging Poly Pail
Vapor Pressure 1mmHg at 440°C
Density 2.165g/cm³
Assay (Dry Basis) >99.5%
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.