missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.

Brand:  Gibco™ A5802903

 View more versions of this product

Product Code. 18064080

  • 2715.00 € / 200 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium Chloride
0.5 M
Liquid
0.5 M NaCl
Sterile-filtered
0.2 μm Sterile-filtered
880–980 mOsm/kg
Clear
4.5–8.0
200 L
Drum
GMP - 21 CFR 820, ISO 13485
≤ 1.0 EU/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.