missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
10 kg
25 kg
32 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 10 kg |
Linear Formula | NaCl |
UN Number | NONH for all modes of transport |
Beilstein | 3534976 |
IUPAC Name | sodium chloride |
Formula Weight | 58.44g/mol |
Percent Purity | 99 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Chemical Name or Material | Sodium Chloride, Meets analytical specification of Ph. Eur.,BP, USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'