missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. 10407152
Click to view available options
Quantity:
1 AMP
6 AMP
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Physical Form Liquid
Quantity 1 AMP
Packaging Ampoule
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'