missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride solution, 0.1M, Honeywell Fluka™

Product Code. 15624920
Click to view available options
Quantity:
500 mL
1 L

Volumetric, 0.1 M NaCl (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Quantity 1 L
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
Formula Weight 58.44g/mol
Concentration or Composition (by Analyte or Components) 0.1M
Chemical Name or Material Sodium chloride solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'