missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, Technical, Rock Salt, Fisher Chemical
Brand: Fisher Chemical S/3100/63
This item is not returnable.
View return policy
Description
3kg, Plastic powder jar
Specifications
1461°C | |
7647-14-5 | |
5.0 to 8.0 | |
3 kg | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium;chloride | |
5234 | |
58.44g/mol | |
1mmHg 865 |
801°C | |
White | |
Solid | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Plastic powder jar |