missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorite, 80%, pure, unstabilized, Thermo Scientific Chemicals

Product Code. 10082870
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium chlorite, 80%, ClNaO2, CAS Number-7758-19-2, 7647-14-5, unii-g538ebv4vf, alcide ld, neo silox d, chlorous acid, sodium salt, caswell no. 755, sodiumchlorite, textone, sodium chlorite, textile, chlorite sodium, 500g, CHEBI:78667, White, 231-836-6, 05,603; 09,423; 11,481; 13,280, 90.44, Pure

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2, 7647-14-5
MDL Number MFCD00003478
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
Synonym sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
PubChem CID 23668197
ChEBI CHEBI:78667
IUPAC Name sodium;chlorite
SMILES [O-]Cl=O.[Na+]

Specifications

Melting Point 180.0°C to 200.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 75% min. (Iodometry)
Molecular Formula ClNaO2
Linear Formula NaClO2
Fieser 05,603; 09,423; 11,481; 13,280
Merck Index 15, 8735
Solubility Information Solubility in water: 390g/L (17°C).
Molecular Weight (g/mol) 90.44
Formula Weight 90.44
Percent Purity 80%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Sodium chlorite, Unstabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium chlorite, unstabilized
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.