missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorite, tech. nominally 80%

Product Code. p-4407876
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11317857

Brand: Thermo Scientific Alfa Aesar 014265.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium chlorite, ClNaO3, CAS Number-7758-19-2, 7775-09-9, 1310-73-2, 497-19-8, 7647-14-5, 7757-82-6, chlorate de sodium, asex, chloric acid, sodium salt, atlacide, tumbleleaf, rasikal, kusatol, chlorsaure, desolet, sodium chlorate, 10kg, White flake/powder, Nominally 80%, CHEBI:65242 | CAS: 7758-19-2 | ClNaO3 | 106.44 g/mol

Sodium chlorite is used in the in situ generation of chlorine dioxide for stripping of textiles, bleaching, pulp and paper industries. It acts as a disinfectant in water treatment plant and as a preservative in eye drops. It is also used in contact lens cleaning solution and for sanitizing air ducts. It is associated with zinc chloride and used as a component in therapeutic rinses, toothpastes, mouth sprays and gels. It is utilized for the synthesis of 4-oxo-2-alkenoic acids from alkyl furans. Further, it is involved as a reagent in Pinnick oxidation reaction to prepare carboxylic acid from aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2, 7775-09-9, 1310-73-2, 497-19-8, 7647-14-5, 7757-82-6
MDL Number MFCD00003478
IUPAC Name sodium;chlorite

Specifications

Melting Point 180°C to 200°C (decomposition)
Color White
Physical Form Flakes or Powder
Quantity 500 g
Assay Percent Range Nominally 80%
Molecular Formula ClNaO3
Synonym sodium;chlorite, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
Solubility Information Soluble in water. Slightly soluble in methanol and ethanol.
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES [Na+].[O-][Cl](=O)=O
Molecular Weight (g/mol) 106.44
PubChem CID 516902
ChEBI CHEBI:65242
Formula Weight 90.44
Grade Technical
Concentration or Composition (by Analyte or Components) Sodium chlorite: 80.0%; Sodium chlorate: 3.0%; Sodium hydroxide: 3.0%; Sodium carbonate: 2.0%; Sodium chloride: 10.0%; Sodium sulphate: 0.3%; Water: 1.7%
Sensitivity Hygroscopic
Appearance White flake/powder
Chemical Name or Material Sodium chlorite
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium chlorite, technical grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Sodium chlorite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.