missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium cyanide, 95+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
Color | White |
Physical Form | Crystalline Powder, Flakes, and Chunks |
Quantity | 5 g |
Assay Percent Range | 95+% |
Linear Formula | NaCN |
Beilstein | 02, IV, 50 |
Fieser | 04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185 |
Merck Index | 15, 8741 |
IUPAC Name | sodium iminomethanide |
Formula Weight | 49 |
Show More |
Safety and Handling
Product Identifier
- Sodium cyanide
- Danger
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'