Learn More
Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
28.70 € - 105.00 €
Identificadores de productos químicos
CAS | 143-33-9 |
---|---|
Molecular Formula | CNNa |
Molecular Weight (g/mol) | 49.01 |
MDL Number | MFCD00003523 |
InChI Key | MNWBNISUBARLIT-UHFFFAOYSA-N |
Synonym | sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h |
PubChem CID | 8929 |
SMILES | [Na+].[C-]#N |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
10497782
|
Thermo Scientific Acros
370310050 |
5 g | Glass bottle |
28.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10018921
|
Thermo Scientific Acros
370310010 |
1 kg | Glass bottle |
105.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentIdentificadores de productos químicos
143-33-9 | |
49.01 | |
MNWBNISUBARLIT-UHFFFAOYSA-N | |
8929 |
CNNa | |
MFCD00003523 | |
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
[Na+].[C-]#N |
Especificaciones
1497.0°C | |
143-33-9 | |
Crystals | |
CNNa | |
MFCD00003523 | |
04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185 | |
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
MNWBNISUBARLIT-UHFFFAOYSA-N | |
sodium;cyanide | |
8929 | |
≥98% | |
Sodium cyanide, 98+% |
562.0°C | |
White | |
98% min. (Argentometry) | |
NaCN | |
02, IV, 50 | |
15, 8741 | |
Solubility in water: 370g/L (20°C). Other solubilities: sparingly soluble in ethanol | |
[Na+].[C-]#N | |
49.01 | |
49 | |
Extra Pure |
Seguridad y manipulación
GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lastin
GHS P Statement
Absorb spillage to prevent material damage.
Wear protective gloves/protective clothing.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-599-4
Recommended Storage : Poison Room
- Sodium cyanide
Palabra de advertencia
- Danger
Clasificación
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
Indicaciones de peligro
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
Consejos de prudencia
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only