missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent

Specifications
Specifications
Melting Point | >242.0°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 95% |
Linear Formula | NaBH3CN |
Fieser | 04,448; 05,607; 06,537; 07,334; 08,454; 09,424; 10,360; 11,481; 12,445; 14,287; 16,305; 17,388 |
Merck Index | 15, 8742 |
Solubility Information | Solubility in water: 2120g/L at 29°C (slow decomposition). Other solubilities: soluble in methanol; soluble in thf,372g/L,at 28°C,410g/L at 46°C,422g/L at 62°C,soluble in diglyme 176g/L at 25°C,slightly soluble in ethanol,insolu |
IUPAC Name | sodium cyanoboranuide |
Show More |
Safety and Handling




Product Identifier
- Sodium cyanoborohydride
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H261-In contact with water releases flammable gases.
- H228-Flammable solid.
- EUH032-Contact with acids liberates very toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction