missing translation for 'onlineSavingsMsg'
Learn More

Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals

58.99 € - 476.37 €

Chemical Identifiers

CAS 145224-92-6
Molecular Formula C24H41NaO5
Molecular Weight (g/mol) 432.58
MDL Number MFCD00150750 MFCD00064139
InChI Key NDVHNZISGVSFMP-WTCAICSISA-M
Synonym 3alpha,5beta,12alpha-3,12-dihydroxycholan-24-oic acid monosodium salt monohydrate
PubChem CID 67096373
SMILES O.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495626
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20759.14
25 g
58.99 €
25g
Please to purchase this item. Need a web account? Register with us today!
11405636
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20759.22
100 g
133.67 €
100g
Please to purchase this item. Need a web account? Register with us today!
11415636
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20759.36
500 g
476.37 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

145224-92-6
432.58
NDVHNZISGVSFMP-WTCAICSISA-M
67096373
C24H41NaO5
MFCD00150750 MFCD00064139
3alpha,5beta,12alpha-3,12-dihydroxycholan-24-oic acid monosodium salt monohydrate
O.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O

Specifications

>300°C
Powder
C24H41NaO5
3643164
Soluble in water.
O.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O
432.58
432.58 (414.58 Anhydrous)
145224-92-6
0.98
MFCD00150750 MFCD00064139
3alpha,5beta,12alpha-3,12-dihydroxycholan-24-oic acid monosodium salt monohydrate
NDVHNZISGVSFMP-WTCAICSISA-M
(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid;sodium;hydrate
67096373
Sodium Deoxycholate Monohydrate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302-H335

missing translation for 'einecsNumber' : 206-132-7

missing translation for 'rtecsNumber' : FZ2250000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Sodium deoxycholate monohydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Sodium deoxycholatemonohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only