missing translation for 'onlineSavingsMsg'
Learn More

Sodium diformamide, 98%, Thermo Scientific Chemicals

42.80 € - 160.00 €

Chemical Identifiers

CAS 18197-26-7
Molecular Formula C2H2NNaO2
Molecular Weight (g/mol) 95.03
MDL Number MFCD00145302
InChI Key QJXDSDLNUKLDBP-UHFFFAOYSA-M
Synonym sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
PubChem CID 10877056
IUPAC Name sodium;diformylazanide
SMILES C(=O)[N-]C=O.[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10321853
View Documents Promotion Details
Thermo Scientific Acros
380430050
5 g Glass bottle
42.80 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10726383
View Documents Promotion Details
Thermo Scientific Acros
380430250
25 g Glass bottle
160.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18197-26-7
95.03
QJXDSDLNUKLDBP-UHFFFAOYSA-M
10877056
C(=O)[N-]C=O.[Na+]
C2H2NNaO2
MFCD00145302
sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
sodium;diformylazanide

Specifications

18197-26-7
Powder
C2H2NNaO2
MFCD00145302
QJXDSDLNUKLDBP-UHFFFAOYSA-M
sodium;diformylazanide
10877056
98%
Sodium diformamide
Beige
98%
NaN(COH)2
sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
C(=O)[N-]C=O.[Na+]
95.03
95.03
Authentic
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only