missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Product Code. 11357327
Click to view available options
Quantity:
250 g
1 kg
5 kg

Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 10049-21-5
Molecular Formula H2NaO4P x H2O
Molecular Weight (g/mol) 137.99
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Especificaciones

Melting Point 100°C (in 2H2O)
Physical Form Crystalline
Quantity 250 g
Merck Index 14,8660
Solubility Information Highly soluble in water. Insoluble in ethanol,ether and chloroform.
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99 (119.98 Anhydrous)
Percent Purity 98.0 to 102.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium dihydrogen phosphate monohydrate
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'