missing translation for 'onlineSavingsMsg'
Learn More
Sodium Dithionite, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10274490

Sodium Dithionite, Extra Pure, SLR, Fisher Chemical™

Product Code. 10274490
500 g
Click to view available options
Quantity:
500 g
3 kg
This item is not returnable. View return policy

Product Code. 10274490

Brand: Fisher Chemical S/3800/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Dithionite, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.096
MDL Number 11640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
pH 8 to 9.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 174.1g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium hydrosulfite
Signal Word
  • Danger
Hazard Category
  • Self-heating substance Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H251-Self-heating: may catch fire.
  • H302-Harmful if swallowed.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Dithionite, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.