Learn More
Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals
Description
Useful chiral synthon; building block for depsipeptides.Sodium DL-lactate, 60% w/w aq. Soln is used as a food additive, plasticizer for casein, preservative, acidity regulator, bulking agent, and in shampoo and liquid soaps. It is also used for the treatment for arrhythmias and sympathomimetics. It is useful chiral synthon and building block for depsipeptides. It is used as a corrosion inhibitor in alcohol antifreeze mixtures, flavoring agent, emulsifier, corrosion inhibitor and pH control agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Density | 1.326 g/mL |
Odor | Odorless |
Linear Formula | C3H5NaO3 |
Quantity | 1 L |
Beilstein | 4332999 |
Merck Index | 14,8635 |
Solubility Information | Miscible with water and alcohol. |
Formula Weight | 112.06 |
Concentration or Composition (by Analyte or Components) | 60% w/w aq. soln. |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.