missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
42.65 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
200 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Description
- DNase, RNase, and protease-free premixed solution containing 10% SDS for protein extraction and DNA extraction procedures
- Reduce preparation time by using this premade SDS solution
- SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
- SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
- Intended for research use only; not for human or animal diagnostic purposes
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Absorbance | 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm |
Color | Colorless |
Physical Form | Liquid |
Boiling Point | 100°C |
Density | 1.01g/cm³ |
Concentration | 9.5 to 10.5% |
Linear Formula | CH3(CH2)10CH2OSO3Na |
Quantity | 1 L |
Merck Index | 15, 8768 |
IUPAC Name | sodium;dodecyl sulfate |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Sodium dodecyl sulfate, 10% solution
- Warning
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'