missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, 95%, MP Biomedicals™
Approximately 95% alkyl sulfate, predominantly composed of C12 and C14 homologs
63.80 € - 125.00 €
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | SDS, Lauryl sulfate sodium salt |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
IUPAC Name | sodium dodecyl sulfate |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11456051
|
MP Biomedicals
0219052290 |
500 g |
63.80 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11466051
|
MP Biomedicals
0219052291 |
1 kg |
125.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
sodium dodecyl sulfate |
C12H25NaO4S | |
MFCD00036175 | |
SDS, Lauryl sulfate sodium salt | |
CHEBI:8984 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
White | |
Granular Powder | |
0.95 | |
MFCD00036175 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
3423265 | |
288.38 |
151-21-3 | |
>1.1 at 20°C (water = 1) | |
C12H25NaO4S | |
SDS, Lauryl sulfate sodium salt | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
Sodium dodecyl sulfate |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium Dodecyl Sulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Dodecyl Sulfate