missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate, 99+%, for Electrophoresis, Fisher Chemical™
72.70 € - 1725.00 €
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Description
Sodium Dodecyl Sulfate, 99+%, for Electrophoresis
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
C12H25NaO4S | |
MFCD00036175 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
CHEBI:8984 |
Specifications
206°C | |
White | |
Solid | |
MFCD00036175 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
3423265 | |
288.38g/mol |
151-21-3 | |
8.5 to 10 | |
C12H25NaO4S | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dodecyl sulfate sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title