missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents

Sodium Dodecyl Sulfate is the most commonly used detergent in protein purification and electrophoresis

66.95 € - 600.00 €

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15440685
View Documents
Fisher BioReagents
BP8200-100
100 g Poly Bottle
66.95 €
100g
Please to purchase this item. Need a web account? Register with us today!
15450685
View Documents
Fisher BioReagents
BP8200-500
500 g Poly Bottle
101.96 €
500g
Please to purchase this item. Need a web account? Register with us today!
15480685
View Documents
Fisher BioReagents
BP8200-5
5 kg Poly Pail
600.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
CHEBI:8984
C12H25NaO4S
MFCD00036175
3423265
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

≤0.10 (0.1% Solution) at 260nm
151-21-3
204°C
Pass test
C12H25NaO4S
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
≥98.0% (C12 alcohol)
Sodium Dodecyl Sulfate (SDS), Micropellets
White
Solid
8.5 to 10
>99%
MFCD00036175
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
Molecular Biology
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature

Product Identifier
  • Dodecyl sulfate sodium salt

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.