missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecylsulfate, 10% Solution, Ultrapure, Thermo Scientific Chemicals

Product Code. 15825428
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Solution
Concentration 10% Solution
Quantity 100 mL
Packaging Nalgene bottle
IUPAC Name sodium dodecyl sulfate
Chemical Name or Material Sodium Dodecylsulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'