missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecylsulfate, ≥98%, Ultrapure, Thermo Scientific Chemicals

Product Code. 15895358
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium;dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Crystalline Flakes
Melting Point ∼206°C
Quantity 1 kg
Packaging Plastic bottle
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
Formula Weight 288.38
Percent Purity ≥98%
Grade Ultra Pure
Chemical Name or Material Sodium Dodecylsulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'