missing translation for 'onlineSavingsMsg'
Learn More

Sodium ethoxide, 21% in ethanol, AcroSeal™, Thermo Scientific Chemicals

23.92 € - 95.07 €

Chemical Identifiers

CAS 141-52-6, 64-17-5
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.04
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10568505
View Documents
Thermo Scientific Acros
429301000
100 mL AcroSeal™ Glass Bottle
23.92 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10568045
View Documents
Thermo Scientific Acros
429308000
800 mL AcroSeal™ Glass Bottle
95.07 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-52-6, 64-17-5
68.04
QDRKDTQENPPHOJ-UHFFFAOYSA-N
2723922
sodium;ethanolate
C2H5NaO
MFCD00012417
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
CHEBI:52096
CC[O-].[Na+]

Specifications

91.0°C
C2H5NaO
MFCD00012417
15,8749
Solubility in water: miscible (hydrolyse)
CC[O-].[Na+]
68.04
CHEBI:52096
1.385
0.8680g/mL
141-52-6, 64-17-5
C2H5ONa
25°C
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
QDRKDTQENPPHOJ-UHFFFAOYSA-N
sodium;ethanolate
2723922
68.04
0.868
Sodium ethoxide, 21% in ethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 205-487-5

Product Identifier
  • Sodium ethoxide, 21% in ethanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

For Manufacturing and Laboratory Use. Not for beverage use