missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride, 99.0+%, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
Description
Sodium Fluoride, 99.0+%, Certified AR for Analysis
Specifications
Specifications
Boiling Point | 1700°C |
Melting Point | 993°C |
Color | White |
pH | 7.4 |
Physical Form | Powder/Solid |
Quantity | 3 kg |
IUPAC Name | sodium fluoride |
Formula Weight | 41.98g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1mmHg 1077 |
Safety and Handling
Product Identifier
- Sodium fluoride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'