missing translation for 'onlineSavingsMsg'
Learn More

Sodium formate, ACS reagent, Thermo Scientific Chemicals

Product Code. 12347183
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.01
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 258.0°C to 261.0°C
Boiling Point 360.0°C
Packaging Plastic bottle
Linear Formula HCO2Na
Quantity 2.5 kg
Merck Index 158756
Solubility Information Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,insoluble in ether
Formula Weight 68.01
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Sodium formate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'